N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

F1082-1G F1082-5G
TCF1082-1GEA 37 USD
TCF1082-1G TCF1082-5G
N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine ≥97.0% (by HPLC, titration analysis)
N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine
Formula: C₁₃H₁₂FN₃O₂
MDL Number: MFCD12828054
CAS Number: 150812-21-8

Order Now

Specification Test Results

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR