2,6-Dibromopyrazine ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
D4544-1G
D4544-5G
TCD4544-1GEA
39.8
USD
TCD4544-1G
TCD4544-5G
2,6-Dibromopyrazine ≥97.0% (by GC)
2,6-Dibromopyrazine
Formula:
C₄H₂Br₂N₂ Boiling Pt: 65°C Storage Temperature: Refrigerator |
MDL Number:
MFCD08061601 CAS Number: 23229-25-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...