Diisopropylamine dichloroacetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Diisopropylammonium dichloroacetate

D5074-1G D5074-5G
TCD5074-1GEA 21 USD
TCD5074-1G TCD5074-5G
Diisopropylamine dichloroacetate ≥98.0% (by titrimetric analysis)
Diisopropylamine dichloroacetate
Formula: C₈H₁₇Cl₂NO₂
MW: 230.129 g/mol
MDL Number: MFCD00868288
CAS Number: 660-27-5
EINECS: 211-538-2
Merck Index: 14,03197

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR