Methyl-2,6-difluoro-3-nitrobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 2,6-difluoro-3-nitrobenzoate

M2749-1G M2749-5G
TCM2749-1GEA 25 USD
TCM2749-1G TCM2749-5G
Methyl-2,6-difluoro-3-nitrobenzoate ≥98.0% (by GC)
Methyl-2,6-difluoro-3-nitrobenzoate
Formula: C₈H₅F₂NO₄
MW: 217.128 g/mol
Boiling Pt: 151°C
MDL Number: MFCD07368850
CAS Number: 84832-01-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR