potassium trimethylacetate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,2-Dimethylpropionic acid potassium salt, Pivalic acid potassium salt, Potassium trimethylacetate, Potassium Pivalate, Trimethylacetic acid potassium salt, Potassium 2,2-Dimethylpropionate
P2354-25G
P2354-5G
TCP2354-25GEA
98
USD
TCP2354-25G
TCP2354-5G
potassium trimethylacetate ≥98.0% (by titrimetric analysis)
potassium trimethylacetate
Formula:
C₅H₉KO₂ MW: 140.223 g/mol |
MDL Number:
MFCD00671345 CAS Number: 19455-23-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...