Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine ≥97.0% (by GC)

Supplier: TCI America

D5135-1G D5135-200MG
TCD5135-1GEA 123 USD
TCD5135-1G TCD5135-200MG
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine ≥97.0% (by GC)
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine
Formula: C₂₇H₄₅P
MDL Number: MFCD11656073
CAS Number: 303111-96-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 137.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR