1-Acetyl-5-bromo-4-chloro-1H-indol-3-ylacetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Acetyl-5-bromo-4-chloro-1H-indol-3-yl acetate, 1-Acetyl-5-bromo-4-chloro-3-indolyl acetate, 3-Acetoxy-1-acetyl-5-bromo-4-chloroindole

A2923-1G A2923-5G
76218-750EA 80 USD
76218-750 76218-752
1-Acetyl-5-bromo-4-chloro-1H-indol-3-ylacetate ≥98.0% (by GC)
1-Acetyl-5-bromo-4-chloro-1H-indol-3-ylacetate
Formula: C₁₂H₉BrClNO₃
MW: 330.562 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00040632
CAS Number: 3030-06-6
EINECS: 221-200-6

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR