Fmoc-L-4-methylphe ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methyl-L-phenylalanine, N-Fmoc-4-methyl-L-phenylalanine, Fmoc-Phe(4-Me)-OH, Fmoc-p-Me-Phe-OH
F1032-1G
F1032-5G
TCF1032-1GEA
58
USD
TCF1032-1G
TCF1032-5G
Fmoc-L-4-methylphe ≥98.0% (by HPLC, titration analysis)
Fmoc-L-4-methylphe
Formula:
C₂₅H₂₃NO₄ MW: 401.462 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00270197 CAS Number: 199006-54-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Specific rotation [a]20/D | -31.0 to -34.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...