Fmoc-GABA-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Fmoc-amino)butyric acid, N-Fmoc-4-aminobutyric acid, Fmoc-γ-Abu-OH, Fmoc-GABA-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric acid

F0911-25G F0911-5G
TCF0911-25GEA 131 USD
TCF0911-25G TCF0911-5G
Fmoc-GABA-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-GABA-OH
Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00144889
CAS Number: 116821-47-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR