Fmoc-D-Thr(tBu)-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F1128-1G F1128-5G
TCF1128-1GEA 52 USD
TCF1128-1G TCF1128-5G
Fmoc-D-Thr(tBu)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-D-Thr(tBu)-OH
Formula: C₂₃H₂₇NO₅
MDL Number: MFCD00080292
CAS Number: 138797-71-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Specific rotation [a]20/D -14.0 to -17.0 deg (C=1, EtOAc)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR