Fmoc-Cys(tBu)-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
F0972-1G
F0972-5G
TCF0972-1GEA
19
USD
TCF0972-1G
TCF0972-5G
Fmoc-Cys(tBu)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Cys(tBu)-OH
Formula:
C₂₂H₂₅NO₄S Storage Temperature: Refrigerator |
MDL Number:
MFCD00037130 CAS Number: 67436-13-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 136.0 to 140.0 deg-C |
Specific rotation [a]20/D | -23.0 to -26.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...