Boc-D-Chg-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Boc-D-Chg-OH, N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine, N-Boc-2-cyclohexyl-D-glycine, Boc-cyclohexyl-D-Gly-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid, BOC-D-α-cyclohexylglycine, N-Boc-D-2-cyclohexylglycine

B4975-1G B4975-5G
TCB4975-1GEA 44 USD
TCB4975-1G TCB4975-5G
Boc-D-Chg-OH ≥98.0% (by HPLC, titration analysis)
Boc-D-Chg-OH
Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Melting Pt: 75°C
MDL Number: MFCD00133629
CAS Number: 70491-05-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -12.0 to -9.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR