Boc-Tyr-OBZL ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(S)-Benzyl 2-((tert-butoxycarbonyl)amino)-3-(4-hydroxyphenyl)propanoate, N-Boc-L-tyrosine Benzyl ester, Boc-Tyr-OBzl, N-(tert-Butoxycarbonyl)-L-tyrosine Benzyl ester
B5343-1G
B5343-5G
TCB5343-1GEA
41
USD
TCB5343-1G
TCB5343-5G
Boc-Tyr-OBZL ≥98.0% (by HPLC)
Boc-Tyr-OBZL
Formula:
C₂₁H₂₅NO₅ MW: 371.433 g/mol |
MDL Number:
MFCD00190840 CAS Number: 19391-35-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 126.0 to 130.0 deg-C |
Specific rotation [a]20/D | -8.0 to -12.0 deg (C=1,MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...