Trimethylphenylammonium hydroxide ∼8.5% in methanol

Supplier: TCI America
Danger

T3610-100ML T3610-25ML
TCT3610-100MLEA 126 USD
TCT3610-100ML TCT3610-25ML
Trimethylphenylammonium hydroxide ∼8.5% in methanol
Trimethylphenylammonium hydroxide
Formula: C₉H₁₅NO
Storage Temperature: Refrigerator
MDL Number: MFCD00041899
CAS Number: 1899-02-1
EINECS: 217-592-3
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid to slightly cloudy liquid
Concentration (Neutralization titration) 8.3 to 9.8 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR