N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
Supplier: TCI America
D5126-1G
D5126-5G
TCD5126-1GEA
85
USD
TCD5126-1G
TCD5126-5G
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine
Formula:
C₄₄H₃₂N₂ |
MDL Number:
MFCD03093246 CAS Number: 123847-85-8 |
Specification Test Results
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...