N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation

Supplier: TCI America

B5140-1G B5140-5G
TCB5140-1GEA 204 USD
TCB5140-1G TCB5140-5G
N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine
Formula: C₄₀H₃₆N₂O₂
MW: 576.74 g/mol
MDL Number: MFCD28384132
CAS Number: 169685-34-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR