Inulin

Supplier: TCI America

Synonyms: Dahlin, Alantin, Inulin (by Enzymatic synthesis)

I1067-25G I1067-500G
TCI1067-25GEA 42.72 USD
TCI1067-25G TCI1067-500G
Inulin
Inulin
MDL Number: MFCD00131407
CAS Number: 9005-80-5
EINECS: 232-684-3
Merck Index: 14,05004

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Drying loss max. 10.0 %
Ignition residue (Sulfate) max. 0.5 %
Dextrin and Starch to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR