9,9-Bis(4-hydroxyphenyl)fluorene ≥98.0% (by GC), purified by sublimation

Supplier: TCI America
Warning

Synonyms: 4,4'-(9-Fluorenylidene)diphenol, 4,4'-(9H-Fluorene-9,9-diyl)diphenol

B4834-1G B4834-5G
TCB4834-1GEA 59 USD
TCB4834-1G TCB4834-5G
9,9-Bis(4-hydroxyphenyl)fluorene ≥98.0% (by GC), purified by sublimation
9,9-Bis(4-hydroxyphenyl)fluorene
Formula: C₂₅H₁₈O₂
MW: 350.42 g/mol
MDL Number: MFCD00191392
CAS Number: 3236-71-3
EINECS: 406-950-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 224.0 to 228.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR