4-Chloro-5-methoxypyridazin-3(2H)-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C2972-1G C2972-200MG
TCC2972-1GEA 190 USD
TCC2972-1G TCC2972-200MG
4-Chloro-5-methoxypyridazin-3(2H)-one ≥98.0% (by GC, titration analysis)
4-Chloro-5-methoxypyridazin-3(2H)-one
Formula: C₅H₅ClN₂O₂
MDL Number: MFCD04971392
CAS Number: 63910-43-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR