Isopropyl-3-aminobut-2-enoate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Isopropyl 3-Aminocrotonate, Isopropyl 3-aminobut-2-enoate, 3-Aminocrotonic acid Isopropyl ester, Isopropyl 3-Amino-2-butenoate, 3-Amino-2-butenoic acid Isopropyl ester
I0884-1G
I0884-5G
TCI0884-1GEA
37
USD
TCI0884-1G
TCI0884-5G
Isopropyl-3-aminobut-2-enoate ≥98.0% (by GC, titration analysis)
Isopropyl-3-aminobut-2-enoate
Formula:
C₇H₁₃NO₂ MW: 143.1836 g/mol Boiling Pt: 80°C |
MDL Number:
MFCD00134272 CAS Number: 14205-46-0 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...