2-Aminothiophene-3-carboxamide ≥98.0% (by GC)

Supplier: TCI America
Warning

A3165-1G A3165-5G
TCA3165-1GEA 30 USD
TCA3165-1G TCA3165-5G
2-Aminothiophene-3-carboxamide ≥98.0% (by GC)
2-Aminothiophene-3-carboxamide
Formula: C₅H₆N₂OS
MW: 142.1789 g/mol
MDL Number: MFCD00706455
CAS Number: 14080-51-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR