3-Oxetanamine ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-amino-oxetane, 3-Aminooxetane

A3080-1G A3080-200MG
TCA3080-1GEA 229 USD
TCA3080-1G TCA3080-200MG
3-Oxetanamine ≥97.0% (by GC)
3-Oxetanamine
Formula: C₃H₇NO
MW: 73.09 g/mol
Flash Pt: 54°C
Storage Temperature: Refrigerator
MDL Number: MFCD08544363
CAS Number: 21635-88-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR