5-Chlorothiophene-2-carbonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

C2864-25G C2864-5G
TCC2864-25GEA 87.9 USD
TCC2864-25G TCC2864-5G
5-Chlorothiophene-2-carbonyl chloride ≥98.0% (by GC, titration analysis)
5-Chlorothiophene-2-carbonyl chloride
Formula: C5H2Cl2OS
Boiling Pt: 225°C
MDL Number: MFCD00043886
CAS Number: 42518-98-9
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR