(3-Carbamoylphenyl)boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C2992-1G C2992-5G
TCC2992-1GEA 115 USD
TCC2992-1G TCC2992-5G
(3-Carbamoylphenyl)boronic acid (contains varying amounts of Anhydride)
(3-Carbamoylphenyl)boronic acid
Formula: C₇H₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03411948
CAS Number: 351422-73-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR