(S)-1-(Naphthalen-1-yl)ethanol ≥97.0% (by GC)

Supplier: TCI America

N1063-1G N1063-5G
76220-372EA 67.18 USD
76220-372 76220-374
(S)-1-(Naphthalen-1-yl)ethanol ≥97.0% (by GC)
(S)-1-(Naphthalen-1-yl)ethanol
Formula: C₁₂H₁₂O
Boiling Pt: 165°C
MDL Number: MFCD00077831
CAS Number: 15914-84-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 44.0 to 48.0 deg-C
Specific rotation [a]20/D -75.0 to -80.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR