2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: XPhos, 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl

D5038-1G D5038-5G
TCD5038-1GEA 99 USD
TCD5038-1G TCD5038-5G
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥98.0% (by GC)
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl
Formula: C₃₃H₄₉P
MW: 476.72 g/mol
MDL Number: MFCD04117682
CAS Number: 564483-18-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 186.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR