5-Bromo-2-ethynylpyridine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5179-1G
B5179-200MG
TCB5179-1GEA
414
USD
TCB5179-1G
TCB5179-200MG
5-Bromo-2-ethynylpyridine ≥98.0% (by GC)
5-Bromo-2-ethynylpyridine
Formula:
C₇H₄BrN |
MDL Number:
MFCD03095276 CAS Number: 111770-86-6 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...