3-(4-Bromophenyl)pyridine ≥96.0% (by GC)

Supplier: TCI America
Warning

B4617-1G B4617-5G
TCB4617-1GEA 54 USD
TCB4617-1G TCB4617-5G
3-(4-Bromophenyl)pyridine ≥96.0% (by GC)
3-(4-Bromophenyl)pyridine
Formula: C₁₁H₈BrN
Storage Temperature: Refrigerator
MDL Number: MFCD04116232
CAS Number: 129013-83-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR