2-Cyano-5-fluorobenzyl bromide ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2-Bromomethyl-4-fluorobenzonitrile

B4988-1G B4988-5G
TCB4988-1GEA 44 USD
TCB4988-1G TCB4988-5G
2-Cyano-5-fluorobenzyl bromide ≥98.0% (by GC)
2-Cyano-5-fluorobenzyl bromide
Formula: C₈H₅BrFN
MDL Number: MFCD08059542
CAS Number: 421552-12-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR