(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride ≥96.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
(H-Cystine-OMe)₂·2HCl, L-Cystine dimethyl ester dihydrochloride, (H-Cys-OMe)₂·2 HCl, (H-Cys-OMe)2.2HCl
C3303-25G
C3303-5G
TCC3303-25GEA
126
USD
TCC3303-25G
TCC3303-5G
(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride ≥96.0% (by total nitrogen and titration analysis)
(2R,2'R)-Dimethyl-3,3'-disulfanediylbis(2-aminopropanoate) dihydrochloride
Formula:
C₈H₁₈Cl₂N₂O₄S₂ MW: 341.28 g/mol |
MDL Number:
MFCD00012490 CAS Number: 32854-09-4 EINECS: 251-261-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 96.0 % |
Purity (Argentmetric Titration) | min. 96.0 % |
Specific rotation [a]20/D | -39.0 to -43.0 deg (C=2,MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...