(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-tert-Butyl L-Aspartate, H-Asp(OtBu)-OH, L-Aspartic acid 4-tert-butyl ester, L-Aspartic acid 4-tert-Butyl ester

B3919-1G B3919-5G
TCB3919-1GEA 67 USD
TCB3919-1G TCB3919-5G
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0% (by titrimetric analysis)
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid
Formula: C₈H₁₅NO₄
MDL Number: MFCD00038577
CAS Number: 3057-74-7
EINECS: 221-292-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +8.0 to +10.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR