2,4-Dichloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)

Supplier: TCI America
Warning

D4805-1G D4805-5G
76219-700EA 19 USD
76219-700 76219-702
2,4-Dichloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)
2,4-Dichloro-6-phenyl-1,3,5-triazine
Formula: C₉H₅Cl₂N₃
Boiling Pt: 136°C
MDL Number: MFCD00047340
CAS Number: 1700-02-3
EINECS: 216-928-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR