Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-Methyl-1,3,4-oxadiazole-2-carboxylic acid potassium salt, Potassium 5-methyl-1,3,4-oxadiazole-2-carboxylate

P2220-1G P2220-200MG
TCP2220-1GEA 356 USD
TCP2220-1G TCP2220-200MG
Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate ≥98.0% (by HPLC, titration analysis)
Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate
Formula: C4H3KN2O3
MW: 166.1800 g/mol
MDL Number: MFCD08060085
CAS Number: 888504-28-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR