2,6-Diiodopyridine ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
D5268-1G
D5268-5G
76219-266EA
165
USD
76219-266
76219-268
2,6-Diiodopyridine ≥97.0% (by GC)
2,6-Diiodopyridine
Formula:
C₅H₃I₂N MW: 330.89 g/mol |
MDL Number:
MFCD07366728 CAS Number: 53710-17-1 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...