3-(4-Bromophenyl)propan-1-ol ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3-(4-Bromophenyl)propyl alcohol, 3-(4-Bromophenyl)-1-propanol
B4657-1G
B4657-5G
TCB4657-1GEA
36
USD
TCB4657-1G
TCB4657-5G
3-(4-Bromophenyl)propan-1-ol ≥98.0% (by GC)
3-(4-Bromophenyl)propan-1-ol
Formula:
C₉H₁₁BrO MW: 215.09 g/mol Boiling Pt: 162°C |
MDL Number:
MFCD09028724 CAS Number: 25574-11-2 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...