Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Phenyl (4,6-Dimethoxy-2-pyrimidinyl)carbamate, (4,6-Dimethoxy-2-pyrimidinyl)carbamic acid Phenyl ester, Phenyl (4,6-dimethoxypyrimidin-2-yl)carbamate

P2182-25G P2182-5G
TCP2182-25GEA 110 USD
TCP2182-25G TCP2182-5G
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate ≥98.0% (by GC, titration analysis)
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Formula: C13H13N3O4
MW: 275.2600 g/mol
MDL Number: MFCD02947731
CAS Number: 89392-03-0
EINECS: 406-600-2
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR