1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Propylpyrazole-4-boronic acid Pinacol ester, 2-(1-Propyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Propyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
P2277-1G
P2277-5G
TCP2277-1GEA
71.1
USD
TCP2277-1G
TCP2277-5G
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula:
C₁₂H₂₁BN₂O₂ Boiling Pt: 102°C |
MDL Number:
MFCD05663871 CAS Number: 827614-69-7 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...