1-Amino-2-(isopropylsulfonyl)benzene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-(Isopropylsulphonyl)aniline, 2-Aminophenyl Isopropyl sulphone
I1033-1G
I1033-5G
76220-180EA
48
USD
76220-180
76220-182
1-Amino-2-(isopropylsulfonyl)benzene ≥98.0% (by GC)
1-Amino-2-(isopropylsulfonyl)benzene
Formula:
C₉H₁₃NO₂S MW: 199.2700 g/mol |
MDL Number:
MFCD08445603 CAS Number: 76697-50-2 |
Specification Test Results
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...