1-Benzhydrylazetidin-3-one ≥96.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Benzhydryl-3-azetidinone, 1-Diphenylmethyl-3-azetidinone
B4799-1G
B4799-5G
TCB4799-1GEA
60
USD
TCB4799-1G
TCB4799-5G
1-Benzhydrylazetidin-3-one ≥96.0% (by GC, titration analysis)
1-Benzhydrylazetidin-3-one
Formula:
C₁₆H₁₅NO MW: 237.30 g/mol |
MDL Number:
MFCD00964539 CAS Number: 40320-60-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 76.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...