2-Methyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2626-1G M2626-5G
TCM2626-1GEA 95 USD
TCM2626-1G TCM2626-5G
2-Methyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Methyl-5-thiazolecarboxylic acid
Formula: C₅H₅NO₂S
MW: 143.1637 g/mol
MDL Number: MFCD01760300
CAS Number: 40004-69-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR