4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol, 4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol, 4-Hydroxy-2-mercapto-6-(trifluoromethyl)pyrimidine
M2739-1G
M2739-5G
TCM2739-1GEA
48
USD
TCM2739-1G
TCM2739-5G
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol ≥98.0% (by HPLC, titration analysis)
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol
Formula:
C₅H₃F₃N₂OS MW: 196.10 g/mol |
MDL Number:
MFCD00112978 CAS Number: 368-54-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 245.0 to 249.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...