Ethyl-1-methyl-1H-imidazole-2-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Methylimidazole-2-carboxylic acid ethyl ester, Ethyl 1-methylimidazole-2-carboxylate, Ethyl 1-methyl-1H-imidazole-2-carboxylate, Ethyl 1-Methylimidazole-2-carboxylate
E1114-1G
E1114-5G
E1114-25G
TCE1114-1GEA
27
USD
TCE1114-1G
TCE1114-5G
TCE1114-25G
Ethyl-1-methyl-1H-imidazole-2-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-1-methyl-1H-imidazole-2-carboxylate
Formula:
C₇H₁₀N₂O₂ MW: 154.17 g/mol Boiling Pt: 110°C |
MDL Number:
MFCD02683326 CAS Number: 30148-21-1 |
Specification Test Results
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 41.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...