(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B4819-1G B4819-5G
TCB4819-1GEA 82 USD
TCB4819-1G TCB4819-5G
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid ≥95.0% (by GC, titration analysis)
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid
Formula: C₄H₇BrO₃
MDL Number: MFCD09751022
CAS Number: 261904-39-6
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 113.0 to 117.0 deg-C
Specific rotation [a]20/D +10.0 to +12.0 deg (C=2.6, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR