5,6-Dichloro-1H-indole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5,6-Dichloroindole

D4924-1G D4924-5G
TCD4924-1GEA 247 USD
TCD4924-1G TCD4924-5G
5,6-Dichloro-1H-indole ≥98.0% (by GC)
5,6-Dichloro-1H-indole
Formula: C₈H₅Cl₂N
MW: 186.0400 g/mol
MDL Number: MFCD01075036
CAS Number: 121859-57-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR