4-Ethynylpyridine hydrochloride ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

E1043-1G E1043-5G
TCE1043-1GEA 77 USD
TCE1043-1G TCE1043-5G
4-Ethynylpyridine hydrochloride ≥95.0% (by HPLC, titration analysis)
4-Ethynylpyridine hydrochloride
Formula: C₇H₆ClN
MW: 139.58 g/mol
MDL Number: MFCD03093912
CAS Number: 352530-29-1

Order Now

Specification Test Results

Appearance Yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR