tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A2893-1G
A2893-5G
TCA2893-1GEA
55
USD
TCA2893-1G
TCA2893-5G
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate ≥98.0% (by titrimetric analysis)
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate
Formula:
C₁₁H₂₂N₂O₂ |
MDL Number:
MFCD01317792 CAS Number: 162167-97-7 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...