2-Cyanoisonicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Cyano-4-pyridinecarboxylic acid
C3396-1G
C3396-200MG
TCC3396-1GEA
235
USD
TCC3396-1G
TCC3396-200MG
2-Cyanoisonicotinic acid ≥98.0% (by GC, titration analysis)
2-Cyanoisonicotinic acid
Formula:
C₇H₄N₂O₂ MW: 148.1200 g/mol |
MDL Number:
MFCD03411729 CAS Number: 161233-97-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...