2-Boc-aminothiazole-5-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Boc-Aminothiazole-5-carboxylic acid, 2-(tert-Butoxycarbonylamino)thiazole-5-carboxylic acid, 2-(Boc-amino)thiazole-5-carboxylic acid

B4552-1G B4552-200MG
76219-406EA 464 USD
76219-406 76219-408
2-Boc-aminothiazole-5-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2-Boc-aminothiazole-5-carboxylic acid
Formula: C₉H₁₂N₂O₄S
MW: 244.2676 g/mol
MDL Number: MFCD06796614
CAS Number: 302964-02-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR