4,5-Diamino-6-hydroxypyrimidine ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5,6-Diamino-4-hydroxypyrimidine, 5,6-Diamino-4-pyrimidinol, 5,6-Diamino-4(1H)-pyrimidinone

D4488-1G D4488-200MG
TCD4488-1GEA 154 USD
TCD4488-1G TCD4488-200MG
4,5-Diamino-6-hydroxypyrimidine ≥96.0% (by HPLC, titration analysis)
4,5-Diamino-6-hydroxypyrimidine
Formula: C₄H₆N₄O
MW: 126.1200 g/mol
MDL Number: MFCD00006114
CAS Number: 1672-50-0

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR