Methyl-1-boc-3-pyrrolidinecarboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Pyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Methyl ester, 1-Boc-3-pyrrolidinecarboxylic acid methyll ester, 1-tert-Butyl 3-Methyl Pyrrolidine-1,3-dicarboxylate, 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid methyll ester, Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate, Methyl 1-Boc-3-pyrrolidinecarboxylate

M2723-1G M2723-5G
TCM2723-1GEA 99 USD
TCM2723-1G TCM2723-5G
Methyl-1-boc-3-pyrrolidinecarboxylate ≥98.0% (by GC)
Methyl-1-boc-3-pyrrolidinecarboxylate
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD04038683
CAS Number: 122684-33-7

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR